missing translation for 'onlineSavingsMsg'
Learn More

5-Chloropyrazine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. C30021G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C30021G Supplier TCI America Supplier No. C30021G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36070-80-1
Molecular Formula C5H3ClN2O2
Molecular Weight (g/mol) 158.541
MDL Number MFCD09033269
InChI Key FXJOTWLLDJYKAG-UHFFFAOYSA-N
Synonym 5-chloro-2-pyrazinecarboxylic acid, 5-chloro-pyrazine-2-carboxylic acid, 5-chloropyrazinoic acid, pyrazinecarboxylic acid, 5-chloro, 5-chlopopyrazine-2-carboxylic acid, 2-pyrazinecarboxylic acid, 5-chloro, 2-carboxy-5-chloropyrazine, 5-chloro-pyrazine-2-carboxylicacid, 2-carboxy-5-chloro-pyrazine, ksc222q7d
PubChem CID 465472
IUPAC Name 5-chloropyrazine-2-carboxylic acid
SMILES C1=C(N=CC(=N1)Cl)C(=O)O

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 1 g
Formula Weight 158.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloropyrazine-2-carboxylic Acid