missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chlorooxindole 97.0+%, TCI America™
$33.56 - $42.87
Chemical Identifiers
| CAS | 17630-75-0 |
|---|---|
| Molecular Formula | C8H6ClNO |
| Molecular Weight (g/mol) | 167.59 |
| MDL Number | MFCD00191927 |
| InChI Key | WWJLCYHYLZZXBE-UHFFFAOYSA-N |
| Synonym | 5-chlorooxindole, 5-chloroindolin-2-one, 5-chloro-1,3-dihydro-2h-indol-2-one, 5-chloro-2-indolinone, 5-chloro-1,3-dihydro-indol-2-one, 5-chloro-2-oxindole, 2h-indol-2-one, 5-chloro-1,3-dihydro, 5-chloro-2,3-dihydro-1h-indol-2-one, 2-indolinone, 5-chloro, 5-chlorooxindol |
| PubChem CID | 152801 |
| IUPAC Name | 5-chloro-2,3-dihydro-1H-indol-2-one |
| SMILES | ClC1=CC=C2NC(=O)CC2=C1 |
Chemical Identifiers
| 17630-75-0 | |
| 167.59 | |
| WWJLCYHYLZZXBE-UHFFFAOYSA-N | |
| 152801 | |
| ClC1=CC=C2NC(=O)CC2=C1 |
| C8H6ClNO | |
| MFCD00191927 | |
| 5-chlorooxindole, 5-chloroindolin-2-one, 5-chloro-1,3-dihydro-2h-indol-2-one, 5-chloro-2-indolinone, 5-chloro-1,3-dihydro-indol-2-one, 5-chloro-2-oxindole, 2h-indol-2-one, 5-chloro-1,3-dihydro, 5-chloro-2,3-dihydro-1h-indol-2-one, 2-indolinone, 5-chloro, 5-chlorooxindol | |
| 5-chloro-2,3-dihydro-1H-indol-2-one |
Specifications
| 17630-75-0 | |
| C8H6ClNO | |
| 1 g | |
| WWJLCYHYLZZXBE-UHFFFAOYSA-N | |
| 5-chloro-2,3-dihydro-1H-indol-2-one | |
| 152801 | |
| ≥97.0% (GC,N) | |
| 5-Chlorooxindole |
| 197°C | |
| MFCD00191927 | |
| 5-chlorooxindole, 5-chloroindolin-2-one, 5-chloro-1,3-dihydro-2h-indol-2-one, 5-chloro-2-indolinone, 5-chloro-1,3-dihydro-indol-2-one, 5-chloro-2-oxindole, 2h-indol-2-one, 5-chloro-1,3-dihydro, 5-chloro-2,3-dihydro-1h-indol-2-one, 2-indolinone, 5-chloro, 5-chlorooxindol | |
| ClC1=CC=C2NC(=O)CC2=C1 | |
| 167.59 | |
| 167.59 | |
| Crystalline Powder |
Safety and Handling
TSCA : No