missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorooxindole 97.0+%, TCI America™

Catalog No. C15091G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15091G 1 g
C15095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15091G Supplier TCI America Supplier No. C15091G
Only null left

Chemical Identifiers

CAS 17630-75-0
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.59
MDL Number MFCD00191927
InChI Key WWJLCYHYLZZXBE-UHFFFAOYSA-N
Synonym 5-chlorooxindole, 5-chloroindolin-2-one, 5-chloro-1,3-dihydro-2h-indol-2-one, 5-chloro-2-indolinone, 5-chloro-1,3-dihydro-indol-2-one, 5-chloro-2-oxindole, 2h-indol-2-one, 5-chloro-1,3-dihydro, 5-chloro-2,3-dihydro-1h-indol-2-one, 2-indolinone, 5-chloro, 5-chlorooxindol
PubChem CID 152801
IUPAC Name 5-chloro-2,3-dihydro-1H-indol-2-one
SMILES ClC1=CC=C2NC(=O)CC2=C1

Specifications

Melting Point 197°C
Quantity 1 g
Formula Weight 167.59
Percent Purity ≥97.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorooxindole