missing translation for 'onlineSavingsMsg'
Learn More

5-Chloronicotinic Acid 95.0+%, TCI America™

Catalog No. C23991G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23991G 1 g
C23995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23991G Supplier TCI America Supplier No. C23991G

Chemical Identifiers

CAS 22620-27-5
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00128901
InChI Key XYLPLVUYPAPCML-UHFFFAOYSA-N
Synonym 5-chloronicotinic acid, 3-pyridinecarboxylic acid, 5-chloro, 5-chloro-nicotinic acid, 5-chloro-3-pyridinecarboxylic acid, 5-chlornicotins, pubchem11089, acmc-209fxn, 3-carboxy-5-chloropyridine, ksc205g2r, 3-pyridinecarboxylicacid, 5-chloro
PubChem CID 247986
IUPAC Name 5-chloropyridine-3-carboxylic acid
SMILES C1=C(C=NC=C1Cl)C(=O)O

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 1 g
Formula Weight 157.55
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloronicotinic Acid