missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)uracil, 97%, Thermo Scientific™

Chemical Identifiers

CAS 3590-48-5
Molecular Formula C5H5ClN2O2
Molecular Weight (g/mol) 160.56
InChI Key UCDUBKRXOPMNGH-UHFFFAOYSA-N
Synonym 5-chloromethyl pyrimidine-2,4 1h,3h-dione, 5-chloromethyl-1,3-dihydropyrimidine-2,4-dione, 2,4 1h,3h-pyrimidinedione, 5-chloromethyl, 5-chloromethyluracil, 5-chloromethyl uracil, acmc-1ady4, 5-chloromethyl-2,4-pyrimidinediol, 5-chloromethyl pyrimidine-2,4-diol, 5-chloromethyl uracil 250mg
PubChem CID 236125
IUPAC Name 5-(chloromethyl)-1H-pyrimidine-2,4-dione
SMILES C1=C(C(=O)NC(=O)N1)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC446190010
SDS View Documents
Thermo Scientific Chemicals
446190010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC446190050
SDS View Documents
Thermo Scientific Chemicals
446190050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3590-48-5
160.56
5-chloromethyl pyrimidine-2,4 1h,3h-dione, 5-chloromethyl-1,3-dihydropyrimidine-2,4-dione, 2,4 1h,3h-pyrimidinedione, 5-chloromethyl, 5-chloromethyluracil, 5-chloromethyl uracil, acmc-1ady4, 5-chloromethyl-2,4-pyrimidinediol, 5-chloromethyl pyrimidine-2,4-diol, 5-chloromethyl uracil 250mg
5-(chloromethyl)-1H-pyrimidine-2,4-dione
C5H5ClN2O2
UCDUBKRXOPMNGH-UHFFFAOYSA-N
236125
C1=C(C(=O)NC(=O)N1)CCl

Specifications

3590-48-5
Glass Bottle
1g
UCDUBKRXOPMNGH-UHFFFAOYSA-N
5-(chloromethyl)-1H-pyrimidine-2,4-dione
236125
97%
96% min. (HPLC)
C5H5ClN2O2
5-chloromethyl pyrimidine-2,4 1h,3h-dione, 5-chloromethyl-1,3-dihydropyrimidine-2,4-dione, 2,4 1h,3h-pyrimidinedione, 5-chloromethyl, 5-chloromethyluracil, 5-chloromethyl uracil, acmc-1ady4, 5-chloromethyl-2,4-pyrimidinediol, 5-chloromethyl pyrimidine-2,4-diol, 5-chloromethyl uracil 250mg
C1=C(C(=O)NC(=O)N1)CCl
160.56
160.56
5-(Chloromethyl)uracil
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only