missing translation for 'onlineSavingsMsg'
Learn More

5-(Chloromethyl)-1-methyl-3-thien-2-yl-1H-pyrazole, ≥97%, Thermo Scientific™

Catalog No. CC47224CB
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC47224CB 250mg
CC47224DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC47224CB Supplier Thermo Scientific Chemicals Supplier No. CC47224CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 876316-96-0
Molecular Formula C9H9ClN2S
Molecular Weight (g/mol) 212.695
MDL Number MFCD08435886
InChI Key GQMRXNSYTBKJTB-UHFFFAOYSA-N
Synonym 5-chloromethyl-1-methyl-3-thien-2-yl-1h-pyrazole, 5-chloromethyl-1-methyl-3-thiophen-2-yl pyrazole, 1h-pyrazole,5-chloromethyl-1-methyl-3-2-thienyl, 2-5-chloromethyl-1-methylpyrazol-3-yl thiophene, 5-chloromethyl-1-methyl-3-thiophen-2-yl-1h-pyrazole
PubChem CID 18525812
IUPAC Name 5-(chloromethyl)-1-methyl-3-thiophen-2-ylpyrazole
SMILES CN1C(=CC(=N1)C2=CC=CS2)CCl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250mg
Formula Weight 212.7
Percent Purity 97%
Chemical Name or Material 5-(Chloromethyl)-1-methyl-3-thien-2-yl-1H-pyrazole
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.