missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroisatoic Anhydride 98.0+%, TCI America™

Catalog No. C27435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27435G 5 g
C274325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27435G Supplier TCI America Supplier No. C27435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4743-17-3
Molecular Formula C8H4ClNO3
Molecular Weight (g/mol) 197.574
MDL Number MFCD00006701
InChI Key MYQFJMYJVJRSGP-UHFFFAOYSA-N
Synonym 6-Chloro-1,2-dihydro -4H-3,1-benzoxazine-2,4-dione
PubChem CID 78480
IUPAC Name 6-chloro-1H-3,1-benzoxazine-2,4-dione
SMILES C1=CC2=C(C=C1Cl)C(=O)OC(=O)N2

Specifications

Melting Point 272°C
Color White-Yellow
Quantity 5 g
Formula Weight 197.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloroisatoic Anhydride