Learn More
5-Chloroisatin, 98%
CAS: 17630-76-1 | C8H4ClNO2 | 181.58 g/mol
Supplier: Thermo Scientific Chemicals A1312506
Description
5-Chloroisatin is a reagent used in the synthesis of Schiff bases through the reaction with 2-methyl-4-nitroaniline or 4-amino-4,5-dihydro-1H-1,2,3-triazole-5-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Chloroisatin is a reagent used in the synthesis of Schiff bases through the reaction with 2-methyl-4-nitroaniline or 4-amino-4,5-dihydro-1H-1,2,3-triazole-5-one.
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
| 5-Chloroisatin | |
| ∼247°C (decomposition) | |
| MFCD00014567 | |
| 383759 | |
| It is insoluble in water. | |
| ClC1=CC=C2NC(=O)C(=O)C2=C1 | |
| 181.58 | |
| 181.58 |
| 17630-76-1 | |
| C8H4ClNO2 | |
| 5 g | |
| 5-chloroisatin, 5-chloroindoline-2,3-dione, 5-chlorisatin, 1h-indole-2,3-dione, 5-chloro, 5-chloro-2,3-dihydro-1h-indole-2,3-dione, 5-chloro isatin, 5-chlor-2,3-dioxoindolin, 5-chlor-1h-indol-2,3-dion, 2hq, pubchem13607 | |
| XHDJYQWGFIBCEP-UHFFFAOYSA-N | |
| 5-chloro-2,3-dihydro-1H-indole-2,3-dione | |
| 87203 | |
| 98% |
Chemical Identifiers
| 17630-76-1 | |
| 181.58 | |
| XHDJYQWGFIBCEP-UHFFFAOYSA-N | |
| 87203 |
| C8H4ClNO2 | |
| MFCD00014567 | |
| 5-chloroisatin, 5-chloroindoline-2,3-dione, 5-chlorisatin, 1h-indole-2,3-dione, 5-chloro, 5-chloro-2,3-dihydro-1h-indole-2,3-dione, 5-chloro isatin, 5-chlor-2,3-dioxoindolin, 5-chlor-1h-indol-2,3-dion, 2hq, pubchem13607 | |
| ClC1=CC=C2NC(=O)C(=O)C2=C1 |
Safety and Handling
EINECSNumber : 241-614-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only