missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroindole 98.0+%, TCI America™

Catalog No. p-7124797
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C11075G 5 g
C110725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C11075G Supplier TCI America Supplier No. C11075G
Only null left

Chemical Identifiers

CAS 17422-32-1
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00005672
InChI Key MYTGFBZJLDLWQG-UHFFFAOYSA-N
Synonym 5-chloroindole, 1h-indole, 5-chloro, 5-chloro indole, 1h-indole, 5-chloro-9ci, 5-chloroindol, 5-chloro-indole, 3fue, zlchem 215, 5-chlor-1h-indole, indole, 5-chloro
PubChem CID 87110
IUPAC Name 5-chloro-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1Cl

Specifications

Melting Point 72°C
Boiling Point 130°C
Quantity 5 g
Formula Weight 151.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloroindole