missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloroindole 98,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 72°C |
| Boiling Point | 130°C |
| Quantity | 5 g |
| Formula Weight | 151.59 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Poudre cristalline |
| Chemical Name or Material | 5-Chloroindole |
Sécurité et manipulation
| TSCA | No |