missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroindole, 99%

Catalog No. AC109310010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109310010 1 g Glass bottle
AC109310050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109310010 Supplier Thermo Scientific Chemicals Supplier No. 109310010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17422-32-1 | C8H6ClN | 151.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17422-32-1
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.6
MDL Number MFCD00005672
InChI Key MYTGFBZJLDLWQG-UHFFFAOYSA-N
Synonym 5-chloroindole, 1h-indole, 5-chloro, 5-chloro indole, 1h-indole, 5-chloro-9ci, 5-chloroindol, 5-chloro-indole, 3fue, zlchem 215, 5-chlor-1h-indole, indole, 5-chloro
PubChem CID 87110
IUPAC Name 5-chloro-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 69°C to 72°C
Color Gray-Green to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in ethanol
Formula Weight 151.6
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloroindole, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-448-9

RUO – Research Use Only