missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroindole-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. C18685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18685G 5 g
C186825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18685G Supplier TCI America Supplier No. C18685G

Chemical Identifiers

CAS 10517-21-2
Molecular Formula C9H5ClNO2
Molecular Weight (g/mol) 194.59
MDL Number MFCD00005613
InChI Key FUQOTYRCMBZFOL-UHFFFAOYSA-M
Synonym 5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783
PubChem CID 82693
IUPAC Name 5-chloro-1H-indole-2-carboxylate
SMILES [O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1

Specifications

Melting Point 290°C
Color White-Yellow
Quantity 5 g
Formula Weight 195.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloroindole-2-carboxylic Acid
RTECSNumber NL5998400
TSCA No