missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzo[b]thiophene, 97%, Thermo Scientific Chemicals

Catalog No. p-7053954
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6174806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6174806 Supplier Thermo Scientific Chemicals Supplier No. H6174806

5-Chlorobenzo[b]thiophene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorobenzo[b]thiophene is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 20532-33-6
Molecular Formula C8H5ClS
Molecular Weight (g/mol) 168.638
MDL Number MFCD00055623
InChI Key SNYURIHMNFPQFL-UHFFFAOYSA-N
Synonym 5-chlorobenzothiophene, 5-chlorobenzo b thiophene, 5-chloro-benzo b thiophene, 5-chloro benzothiophene, benzo b thiophene, 5-chloro, zlchem 66, 5-chlorothionaphthene, 5-chloranyl-1-benzothiophene, benzo b thiophene,5-chloro, ksc544i5d
PubChem CID 11309754
IUPAC Name 5-chloro-1-benzothiophene
SMILES C1=CC2=C(C=CS2)C=C1Cl

Specifications

Color White
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 168.64
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Chlorobenzo[b]thiophene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only