missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzo[b]thiophene-3-acetic acid, 97%

Catalog No. p-7046021
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL12836MD 250 mg
AAL1283603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL12836MD Supplier Thermo Scientific Chemicals Supplier No. L12836MD

CAS: 17266-30-7 | C10H7ClO2S | 226.674 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17266-30-7
Molecular Formula C10H7ClO2S
Molecular Weight (g/mol) 226.674
MDL Number MFCD00052308
InChI Key QQKKTOPRRGBBCT-UHFFFAOYSA-N
Synonym 5-chlorobenzo b thiophene-3-acetic acid, 2-5-chlorobenzo b thiophen-3-yl acetic acid, 2-5-chloro-1-benzothiophen-3-yl acetic acid, benzo b thiophene-3-acetic acid, 5-chloro, 5-chloro-1-benzothien-3-yl acetic acid, 5-chloro-1-benzothiophen-3-yl acetic acid, maybridge4_002620, acmc-20am45
PubChem CID 205058
IUPAC Name 2-(5-chloro-1-benzothiophen-3-yl)acetic acid
SMILES C1=CC2=C(C=C1Cl)C(=CS2)CC(=O)O

Specifications

Melting Point 152°C to 155°C
Quantity 250 mg
Beilstein 1244795
Formula Weight 226.68
Percent Purity 97%
Chemical Name or Material 5-Chlorobenzo[b]thiophene-3-acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber DL8895000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only