missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzo-2,1,3-thiadiazole, 98%

Catalog No. AAA1828603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1828603 1 g
AAA1828606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1828603 Supplier Thermo Scientific Chemicals Supplier No. A1828603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2207-32-1 | C6H3ClN2S | 170.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2207-32-1
Molecular Formula C6H3ClN2S
Molecular Weight (g/mol) 170.61
MDL Number MFCD00174269
InChI Key VRNJWKISMWDTAY-UHFFFAOYSA-N
Synonym 5-chlorobenzo c 1,2,5 thiadiazole, 5-chlorobenzo-2,1,3-thiadiazole, 2,1,3-benzothiadiazole, 5-chloro, 5-chlorobenzo c 1,2,5-thiadiazole, acmc-1cipd, maybridge1_004942, 5-chloranyl-2,1,3-benzothiadiazole, 3-chloro-8-thia-7,9-diazabicyclo 4.3.0 nona-2,4,6,9-tetraene
PubChem CID 284123
IUPAC Name 5-chloro-2,1,3-benzothiadiazole
SMILES ClC1=CC2=NSN=C2C=C1

Specifications

Melting Point 55°C to 57°C
Quantity 1 g
Formula Weight 170.62
Percent Purity 98%
Chemical Name or Material 5-Chlorobenzo-2,1,3-thiadiazole
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only