missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-8-hydroxyquinoline 98.0+%, TCI America™

Catalog No. p-7124541
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
C064525G 25 g
C0645250G 250 g
2 options

Catalog No. C064525G

Supplier: TCI America C064525G

Only null left

Chemical Identifiers

CAS 130-16-5
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00006788
InChI Key CTQMJYWDVABFRZ-UHFFFAOYSA-N
Synonym 5-chloro-8-hydroxyquinoline, cloxiquine, cloxyquin, 5-chloro-8-quinolinol, dermofongin, 5-chlorooxine, 8-quinolinol, 5-chloro, chlorisept, dermofungin, 5-chloro-8-oxyquinoline
PubChem CID 2817
IUPAC Name 5-chloroquinolin-8-ol
SMILES C1=CC2=C(C=CC(=C2N=C1)O)Cl

Specifications

Melting Point 127°C
Quantity 25 g
Formula Weight 179.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-8-hydroxyquinoline
EINECSNumber (9)-2220
RTECSNumber VC4590000
TSCA Yes