missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-8-hydroxyquinoline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 127°C |
| Quantity | 250 g |
| Formula Weight | 179.60 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 5-Chloro-8-hydroxyquinoline |
Safety and Handling
| EINECSNumber | (9)-2220 |
| RTECSNumber | VC4590000 |
| TSCA | Yes |