missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-8-hydroxyquinoline, 98%

Catalog No. AC109301000
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109301000 100 g Plastic bottle
AC109300050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109301000 Supplier Thermo Scientific Chemicals Supplier No. 109301000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 130-16-5 | C9H6ClNO | 179.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 130-16-5
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.61
MDL Number MFCD00006788
InChI Key CTQMJYWDVABFRZ-UHFFFAOYSA-N
Synonym 5-chloro-8-hydroxyquinoline, cloxiquine, cloxyquin, 5-chloro-8-quinolinol, dermofongin, 5-chlorooxine, 8-quinolinol, 5-chloro, chlorisept, dermofungin, 5-chloro-8-oxyquinoline
PubChem CID 2817
IUPAC Name 5-chloroquinolin-8-ol
SMILES C1=CC2=C(C=CC(=C2N=C1)O)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 121°C to 126°C
Color Gray to Green
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Plastic bottle
Quantity 100 g
Beilstein 21, 95
Merck Index 15, 2405
Formula Weight 179.61
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Chloro-8-hydroxyquinoline, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-978-1
RTECSNumber VC4590000
TSCA TSCA

RUO – Research Use Only