missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-4-nitrothiophene-2-sulfonyl chloride, 98%

Catalog No. AAA1177903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1177903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1177903 Supplier Thermo Scientific Chemicals Supplier No. A1177903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58457-24-2 | C4HCl2NO4S2 | 262.075 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58457-24-2
Molecular Formula C4HCl2NO4S2
Molecular Weight (g/mol) 262.075
MDL Number MFCD00067978
InChI Key SGHAHFHTASPNKN-UHFFFAOYSA-N
Synonym 2-chloro-3-nitrothiophene-5-sulphonyl chloride, 2-chloro-3-nitrothiophene-5-sulfonyl chloride, 5-chloro-4-nitro-thiophene-2-sulfonyl chloride, chloro 5-chloro-4-nitro 2-thienyl sulfone, pubchem5498, 5-chloro-4-nitrothiophene-2-sulfonylchloride, acmc-20am48, 2-chlorosulphonyl-4-nitro-5-chlorothiophene, 5-chloro-4-nitro-2-thienylsulfonyl chloride
PubChem CID 2735783
IUPAC Name 5-chloro-4-nitrothiophene-2-sulfonyl chloride
SMILES C1=C(SC(=C1[N+](=O)[O-])Cl)S(=O)(=O)Cl

Specifications

Melting Point 52°C to 54°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 262.09
Percent Purity 98%
Chemical Name or Material 5-Chloro-4-nitrothiophene-2-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only