missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-4-nitrothiophene-2-sulfonyl chloride, 98%

CAS: 58457-24-2 | C4HCl2NO4S2 | 262.075 g/mol

$71.49 - $71.49

Chemical Identifiers

CAS 58457-24-2
Molecular Formula C4HCl2NO4S2
Molecular Weight (g/mol) 262.075
MDL Number MFCD00067978
InChI Key SGHAHFHTASPNKN-UHFFFAOYSA-N
Synonym 2-chloro-3-nitrothiophene-5-sulphonyl chloride, 2-chloro-3-nitrothiophene-5-sulfonyl chloride, 5-chloro-4-nitro-thiophene-2-sulfonyl chloride, chloro 5-chloro-4-nitro 2-thienyl sulfone, pubchem5498, 5-chloro-4-nitrothiophene-2-sulfonylchloride, acmc-20am48, 2-chlorosulphonyl-4-nitro-5-chlorothiophene, 5-chloro-4-nitro-2-thienylsulfonyl chloride
PubChem CID 2735783
IUPAC Name 5-chloro-4-nitrothiophene-2-sulfonyl chloride
SMILES C1=C(SC(=C1[N+](=O)[O-])Cl)S(=O)(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1177903
View Documents
Thermo Scientific Chemicals
A1177903
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

58457-24-2
262.075
SGHAHFHTASPNKN-UHFFFAOYSA-N
2735783
C1=C(SC(=C1[N+](=O)[O-])Cl)S(=O)(=O)Cl
C4HCl2NO4S2
MFCD00067978
2-chloro-3-nitrothiophene-5-sulphonyl chloride, 2-chloro-3-nitrothiophene-5-sulfonyl chloride, 5-chloro-4-nitro-thiophene-2-sulfonyl chloride, chloro 5-chloro-4-nitro 2-thienyl sulfone, pubchem5498, 5-chloro-4-nitrothiophene-2-sulfonylchloride, acmc-20am48, 2-chlorosulphonyl-4-nitro-5-chlorothiophene, 5-chloro-4-nitro-2-thienylsulfonyl chloride
5-chloro-4-nitrothiophene-2-sulfonyl chloride

Specifications

58457-24-2
C4HCl2NO4S2
1 g
Moisture sensitive
SGHAHFHTASPNKN-UHFFFAOYSA-N
5-chloro-4-nitrothiophene-2-sulfonyl chloride
2735783
98%
52°C to 54°C
MFCD00067978
UN3261
2-chloro-3-nitrothiophene-5-sulphonyl chloride, 2-chloro-3-nitrothiophene-5-sulfonyl chloride, 5-chloro-4-nitro-thiophene-2-sulfonyl chloride, chloro 5-chloro-4-nitro 2-thienyl sulfone, pubchem5498, 5-chloro-4-nitrothiophene-2-sulfonylchloride, acmc-20am48, 2-chlorosulphonyl-4-nitro-5-chlorothiophene, 5-chloro-4-nitro-2-thienylsulfonyl chloride
C1=C(SC(=C1[N+](=O)[O-])Cl)S(=O)(=O)Cl
262.075
262.09
5-Chloro-4-nitrothiophene-2-sulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only