missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-4-fluoro-2-nitroaniline 98.0+%, TCI America™

Catalog No. p-7125851
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31411G 1 g
C31415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31411G Supplier TCI America Supplier No. C31411G
Only null left

Chemical Identifiers

CAS 104222-34-6
Molecular Formula C6H4ClFN2O2
Molecular Weight (g/mol) 190.558
MDL Number MFCD00052698
InChI Key VRJKEIWZSOHDOH-UHFFFAOYSA-N
PubChem CID 517835
IUPAC Name 5-chloro-4-fluoro-2-nitroaniline
SMILES C1=C(C(=CC(=C1Cl)F)[N+](=O)[O-])N

Specifications

Melting Point 142°C
Quantity 1 g
Synonym 2-nitro-4-fluoro-5-chloroaniline, benzenamine, 5-chloro-4-fluoro-2-nitro, benzenamine,5-chloro-4-fluoro-2-nitro, 5-chloro-4-fluoro-2-nitrophenylamine, pubchem8452, acmc-2098bg, timtec-bb sbb003647, buttpark 45\03-11, 3-chloro-4-fluoro-6-nitroaniline, 5-chloro-4-fluoro-2-nitro-aniline
Formula Weight 190.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-4-fluoro-2-nitroaniline