missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-4-fluoro-2-nitroaniline, 97%

Catalog No. AAA1387914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1387914 25 g
AAA1387906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1387914 Supplier Thermo Scientific Chemicals Supplier No. A1387914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 104222-34-6 | C6H4ClFN2O2 | 190.558 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104222-34-6
Molecular Formula C6H4ClFN2O2
Molecular Weight (g/mol) 190.558
MDL Number MFCD00052698
InChI Key VRJKEIWZSOHDOH-UHFFFAOYSA-N
Synonym 2-nitro-4-fluoro-5-chloroaniline, benzenamine, 5-chloro-4-fluoro-2-nitro, benzenamine,5-chloro-4-fluoro-2-nitro, 5-chloro-4-fluoro-2-nitrophenylamine, pubchem8452, acmc-2098bg, timtec-bb sbb003647, buttpark 45\03-11, 3-chloro-4-fluoro-6-nitroaniline, 5-chloro-4-fluoro-2-nitro-aniline
PubChem CID 517835
IUPAC Name 5-chloro-4-fluoro-2-nitroaniline
SMILES C1=C(C(=CC(=C1Cl)F)[N+](=O)[O-])N

Specifications

Melting Point 141°C to 145°C
Quantity 25 g
UN Number UN2811
Beilstein 7202993
Formula Weight 190.56
Percent Purity 97%
Chemical Name or Material 5-Chloro-4-fluoro-2-nitroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only