missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-methyl-1-phenyl-1H-pyrazole, 98%

Catalog No. AAA1268503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1268503 1 g
AAA1268506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1268503 Supplier Thermo Scientific Chemicals Supplier No. A1268503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1131-17-5 | C10H9ClN2 | 192.646 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1131-17-5
Molecular Formula C10H9ClN2
Molecular Weight (g/mol) 192.646
MDL Number MFCD00067853
InChI Key ZZOWFLAMMWOSCG-UHFFFAOYSA-N
Synonym 5-chloro-3-methyl-1-phenyl-1h-pyrazole, 1h-pyrazole, 5-chloro-3-methyl-1-phenyl, buttpark 48\06-20, acmc-20am49, 1-phenyl-3-methyl-5-chloropyrazole, 1-phenyl-3-methyl-5-chloro-1h-pyrazole, 1h-pyrazole,5-chloro-3-methyl-1-phenyl, 5-chloro-3-methyl-1-phenylpyrazole
PubChem CID 2735771
IUPAC Name 5-chloro-3-methyl-1-phenylpyrazole
SMILES CC1=NN(C(=C1)Cl)C2=CC=CC=C2

Specifications

Boiling Point 114°C to 115°C (4 mmHg)
Quantity 1 g
Formula Weight 192.64
Percent Purity 98%
Chemical Name or Material 5-Chloro-3-methyl-1-phenyl-1H-pyrazole
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only