missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-3-hydroxypyridine, 99%

CAS: 74115-12-1 | C5H4ClNO | 129.543 g/mol

Supplier:  Thermo Scientific Chemicals L0756806

 View more versions of this product

Catalog No. AAL0756806


Only null left
Explore available promotions

Description

Description

5-Chloro-3-hydroxypyridine is used in the preparation of 4-(5-chloropyridin-3-yloxy)isoquinoline

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-3-hydroxypyridine is used in the preparation of 4-(5-chloropyridin-3-yloxy)isoquinoline

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

5-Chloro-3-hydroxypyridine
159°C to 163°C
MFCD00006379
1422550
Soluble in water.
C1=C(C=NC=C1Cl)O
129.543
129.55
74115-12-1
C5H4ClNO
5 g
5-chloro-3-hydroxypyridine, 5-chloro-3-pyridinol, 3-chloro-5-hydroxypyridine, 3-pyridinol, 5-chloro, 5chloro-3-pyridinol, pubchem6628, 3-chloro-5-pyridinol, acmc-209otf, 5-chloranylpyridin-3-ol, 3chloro-5-hydroxypyridine
TUIDQYRZDZRHPQ-UHFFFAOYSA-N
5-chloropyridin-3-ol
582305
99%

Chemical Identifiers

74115-12-1
129.543
TUIDQYRZDZRHPQ-UHFFFAOYSA-N
582305
C1=C(C=NC=C1Cl)O
C5H4ClNO
MFCD00006379
5-chloro-3-hydroxypyridine, 5-chloro-3-pyridinol, 3-chloro-5-hydroxypyridine, 3-pyridinol, 5-chloro, 5chloro-3-pyridinol, pubchem6628, 3-chloro-5-pyridinol, acmc-209otf, 5-chloranylpyridin-3-ol, 3chloro-5-hydroxypyridine
5-chloropyridin-3-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 277-727-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only