missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-thiophenecarboxaldehyde, 97%

Catalog No. p-91701
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405080010 1 g Glass bottle
AC405080050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC405080010 Supplier Thermo Scientific Chemicals Supplier No. 405080010
Only null left

CAS: 7283-96-7 | C5H3ClOS | 146.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7283-96-7
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.59
MDL Number MFCD00047090
InChI Key VWYFITBWBRVBSW-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenecarboxaldehyde, 2-chloro-5-thiophenecarboxaldehyde, 5-chlorothiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 5-chloro, 5-chloro-2-formylthiophene, 2-chloro-5-formylthiophene, 5-chlorothiophene-2-aldehyde, 5-chloro-2-thiophenecarbaldehyde, 5-chloro-2-thiophene-carboxaldehyde, pubchem7742
PubChem CID 81700
IUPAC Name 5-chlorothiophene-2-carbaldehyde
SMILES ClC1=CC=C(S1)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Tan to White
Density 1.3760g/mL
Boiling Point 94°C (18.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6040 to 1.606
Quantity 1 g
Specific Gravity 1.376
Solubility Information Solubility in water: insoluble
Formula Weight 146.6
Percent Purity 97%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 5-Chloro-2-thiophenecarboxaldehyde, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air
EINECSNumber 230-708-7
TSCA TSCA

RUO – Research Use Only