missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-pentanone ethylene ketal, 97%

Catalog No. p-90904
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166040050 5 g Glass bottle
AC166040250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC166040050 Supplier Thermo Scientific Chemicals Supplier No. 166040050

CAS: 5978-08-5 | C7H13ClO2 | 164.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5978-08-5
Molecular Formula C7H13ClO2
Molecular Weight (g/mol) 164.63
MDL Number MFCD00003217
InChI Key OFERIRWCHSOJJT-UHFFFAOYSA-N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 73°C to 76°C (7.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4490 to 1.451
Quantity 5 g
Specific Gravity 1.09
Synonym 2-3-chloropropyl-2-methyl-1,3-dioxolane, 5-chloro-2-pentanone ethylene ketal, 2,2-ethylenedioxy-5-chloropentane, 1,3-dioxolane, 2-3-chloropropyl-2-methyl, 5-chloro-2-pentanoneethyleneketal, acmc-20am4f, oferirwchsojjt-uhfffaoysa, 1-chloro-4-pentanone ethylene ketal, 1, 2-3-chloropropyl-2-methyl, 5-chloro-2-pentanone ethylene acetal
SMILES CC1(OCCO1)CCCCl
IUPAC Name 2-(3-chloropropyl)-2-methyl-1,3-dioxolane
PubChem CID 80079
Formula Weight 164.63
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-pentanone ethylene ketal, 97%
Show More Show Less
EINECSNumber 227-776-5

RUO – Research Use Only