missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(methylthio)pyrimidine-4-carboxylic Acid 97.0+%, TCI America™

Catalog No. c25491g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25491G 1 g
C25495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25491G Supplier TCI America Supplier No. C25491G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61727-33-1
Molecular Formula C6H5ClN2O2S
Molecular Weight (g/mol) 204.628
MDL Number MFCD00173907
InChI Key SEPCYCDQJZTPHO-UHFFFAOYSA-N
Synonym 5-chloro-2-methylthio pyrimidine-4-carboxylic acid, 5-chloro-2-methylsulfanyl pyrimidine-4-carboxylic acid, 4-pyrimidinecarboxylic acid, 5-chloro-2-methylthio, 5-chloro-2-methylsulfanyl-pyrimidine-4-carboxylic acid, 4-carboxy-5-chloro-2-methylthio pyrimidine, 5-chloro-2-methylthio-4-pyrimidinecarboxylic acid, 5-chloro-2-methylthiopyrimidine-4-carboxylic acid, zlchem 91, oleosol fast red bl, oleosol fast red gl
PubChem CID 725711
IUPAC Name 5-chloro-2-methylsulfanylpyrimidine-4-carboxylic acid
SMILES CSC1=NC=C(C(=N1)C(=O)O)Cl

Specifications

Melting Point 170°C
Quantity 1 g
Formula Weight 204.63
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-(methylthio)pyrimidine-4-carboxylic Acid