missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2-(methylthio)pyrimidine-4-carboxylic acid, 97%, Thermo Scientific™

Catalog No. p-2701441
Change view
Click to view available options
Quantity:
1g
10g
25g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB10252DA 1g Amber Glass Bottle
BTB10252EA 10g Amber Glass Bottle
BTB10252EB 25g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB10252DA Supplier Thermo Scientific Chemicals Supplier No. BTB10252DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61727-33-1
Molecular Formula C6H5ClN2O2S
Molecular Weight (g/mol) 204.628
MDL Number MFCD00173907
InChI Key SEPCYCDQJZTPHO-UHFFFAOYSA-N
Synonym 5-chloro-2-methylthio pyrimidine-4-carboxylic acid, 5-chloro-2-methylsulfanyl pyrimidine-4-carboxylic acid, 4-pyrimidinecarboxylic acid, 5-chloro-2-methylthio, 5-chloro-2-methylsulfanyl-pyrimidine-4-carboxylic acid, 4-carboxy-5-chloro-2-methylthio pyrimidine, 5-chloro-2-methylthio-4-pyrimidinecarboxylic acid, 5-chloro-2-methylthiopyrimidine-4-carboxylic acid, zlchem 91, oleosol fast red bl, oleosol fast red gl
PubChem CID 725711
IUPAC Name 5-chloro-2-methylsulfanylpyrimidine-4-carboxylic acid
SMILES CSC1=NC=C(C(=N1)C(=O)O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 204.64
Percent Purity 97%
Chemical Name or Material 5-chloro-2-(methylthio)pyrimidine-4-carboxylic acid
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.