missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylindole 98.0+%, TCI America™

Catalog No. C17611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17611G 1 g
C17615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17611G Supplier TCI America Supplier No. C17611G

Chemical Identifiers

CAS 1075-35-0
Molecular Formula C9H8ClN
Molecular Weight (g/mol) 165.62
MDL Number MFCD00005619
InChI Key WUVWAXJXPRYUME-UHFFFAOYSA-N
Synonym 5-chloro-2-methylindole, 2-methyl-5-chloroindole, 1h-indole, 5-chloro-2-methyl, 5-chloro-2-methyl-indole, 2-methyl-5-chloro-1h-indole, pubchem17201, acmc-1bpvo, 5-chlor-2-methyl-1h-indole, indole, 5-chloro-2-methyl, 1h-indole,5-chloro-2-methyl
PubChem CID 70636
IUPAC Name 5-chloro-2-methyl-1H-indole
SMILES CC1=CC2=C(N1)C=CC(=C2)Cl

Specifications

Melting Point 117°C
Boiling Point 140°C
Quantity 1 g
Formula Weight 165.62
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methylindole