missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylindole, 98%

CAS: 1075-35-0 | C9H8ClN | 165.62 g/mol

$72.01 - $241.85

Chemical Identifiers

CAS 1075-35-0
Molecular Formula C9H8ClN
Molecular Weight (g/mol) 165.62
MDL Number MFCD00005619
InChI Key WUVWAXJXPRYUME-UHFFFAOYSA-N
Synonym 5-chloro-2-methylindole, 2-methyl-5-chloroindole, 1h-indole, 5-chloro-2-methyl, 5-chloro-2-methyl-indole, 2-methyl-5-chloro-1h-indole, pubchem17201, acmc-1bpvo, 5-chlor-2-methyl-1h-indole, indole, 5-chloro-2-methyl, 1h-indole,5-chloro-2-methyl
PubChem CID 70636
IUPAC Name 5-chloro-2-methyl-1H-indole
SMILES CC1=CC2=C(N1)C=CC(=C2)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2103603
View Documents
Thermo Scientific Chemicals
B2103603
1 g
Each for $72.01
Only null left
 
AAB2103606
View Documents
Thermo Scientific Chemicals
B2103606
5 g
Each for $241.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1075-35-0
165.62
WUVWAXJXPRYUME-UHFFFAOYSA-N
70636
CC1=CC2=C(N1)C=CC(=C2)Cl
C9H8ClN
MFCD00005619
5-chloro-2-methylindole, 2-methyl-5-chloroindole, 1h-indole, 5-chloro-2-methyl, 5-chloro-2-methyl-indole, 2-methyl-5-chloro-1h-indole, pubchem17201, acmc-1bpvo, 5-chlor-2-methyl-1h-indole, indole, 5-chloro-2-methyl, 1h-indole,5-chloro-2-methyl
5-chloro-2-methyl-1H-indole

Specifications

1075-35-0
C9H8ClN
1 g
WUVWAXJXPRYUME-UHFFFAOYSA-N
5-chloro-2-methyl-1H-indole
70636
98%
110°C to 114°C
MFCD00005619
5-chloro-2-methylindole, 2-methyl-5-chloroindole, 1h-indole, 5-chloro-2-methyl, 5-chloro-2-methyl-indole, 2-methyl-5-chloro-1h-indole, pubchem17201, acmc-1bpvo, 5-chlor-2-methyl-1h-indole, indole, 5-chloro-2-methyl, 1h-indole,5-chloro-2-methyl
CC1=CC2=C(N1)C=CC(=C2)Cl
165.62
165.62
5-Chloro-2-methylindole
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-052-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only