missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylindole, 98%

Catalog No. AAB2103603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2103603 1 g
AAB2103606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2103603 Supplier Thermo Scientific Chemicals Supplier No. B2103603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1075-35-0 | C9H8ClN | 165.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1075-35-0
Molecular Formula C9H8ClN
Molecular Weight (g/mol) 165.62
MDL Number MFCD00005619
InChI Key WUVWAXJXPRYUME-UHFFFAOYSA-N
Synonym 5-chloro-2-methylindole, 2-methyl-5-chloroindole, 1h-indole, 5-chloro-2-methyl, 5-chloro-2-methyl-indole, 2-methyl-5-chloro-1h-indole, pubchem17201, acmc-1bpvo, 5-chlor-2-methyl-1h-indole, indole, 5-chloro-2-methyl, 1h-indole,5-chloro-2-methyl
PubChem CID 70636
IUPAC Name 5-chloro-2-methyl-1H-indole
SMILES CC1=CC2=C(N1)C=CC(=C2)Cl

Specifications

Melting Point 110°C to 114°C
Quantity 1 g
Formula Weight 165.62
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methylindole
EINECSNumber 214-052-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only