missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylbenzenesulfonyl chloride, 97%

Catalog No. AAH3291206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3291206 5 g
AAH3291203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3291206 Supplier Thermo Scientific Chemicals Supplier No. H3291206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34981-38-9 | C7H6Cl2O2S | 225.083 g/mol

5-Chloro-2-methylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-methylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.

Chemical Identifiers

CAS 34981-38-9
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.083
MDL Number MFCD09742738
InChI Key VHBFBNCERXCECQ-UHFFFAOYSA-N
Synonym 4-chlorotoluene-2-sulphonyl chloride, 4-chlorotoluene-2-sulphonylchloride, 2-chlorosulfonyl-4-chlorotoluene, 5-chloro-2-methylbenzene-1-sulfonyl chloride, 5-chloro-2-methylbenzenesulfonylchloride, benzenesulfonyl chloride, 5-chloro-2-methyl, 5-chloro-2-methylbenzensulfonyl chloride, 5-chloro-2-methylphenylsulfonyl chloride, 3-chloro-6-methyl-benzenesulfonyl chloride, benzenesulfonylchloride, 5-chloro-2-methyl
PubChem CID 100415
IUPAC Name 5-chloro-2-methylbenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)Cl)S(=O)(=O)Cl

Specifications

Refractive Index 1.5697
Quantity 5 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 225.09
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-methylbenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 252-306-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only