Learn More
5-Chloro-2-methylaniline, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 95-79-4 |
|---|---|
| Molecular Formula | C7H8ClN |
| Molecular Weight (g/mol) | 141.60 |
| MDL Number | MFCD00007779 |
| InChI Key | WRZOMWDJOLIVQP-UHFFFAOYSA-N |
| Synonym | 5-chloro-o-toluidine, 2-amino-4-chlorotoluene, 2-methyl-5-chloroaniline, fast red kb base, benzenamine, 5-chloro-2-methyl, ansibase red kb, red kb base, 4-chloro-2-aminotoluene, 3-chloro-6-methylaniline, pharmazoid red kb |
| PubChem CID | 7260 |
| ChEBI | CHEBI:82422 |
| IUPAC Name | 5-chloro-2-methylaniline |
| SMILES | CC1=CC=C(Cl)C=C1N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC109380050
|
Thermo Scientific Chemicals
109380050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC109381000
|
Thermo Scientific Chemicals
109381000 |
100g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC109385000
|
Thermo Scientific Chemicals
109385000 |
500g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 95-79-4 | |
| 141.60 | |
| WRZOMWDJOLIVQP-UHFFFAOYSA-N | |
| 7260 | |
| 5-chloro-2-methylaniline |
| C7H8ClN | |
| MFCD00007779 | |
| 5-chloro-o-toluidine, 2-amino-4-chlorotoluene, 2-methyl-5-chloroaniline, fast red kb base, benzenamine, 5-chloro-2-methyl, ansibase red kb, red kb base, 4-chloro-2-aminotoluene, 3-chloro-6-methylaniline, pharmazoid red kb | |
| CHEBI:82422 | |
| CC1=CC=C(Cl)C=C1N |
Specifications
| 95-79-4 | |
| 100.0 | |
| 160°C | |
| 97.5% min. (GC) | |
| C7H8ClN | |
| ClC6H3(CH3)NH2 | |
| 5g | |
| 5-chloro-o-toluidine, 2-amino-4-chlorotoluene, 2-methyl-5-chloroaniline, fast red kb base, benzenamine, 5-chloro-2-methyl, ansibase red kb, red kb base, 4-chloro-2-aminotoluene, 3-chloro-6-methylaniline, pharmazoid red kb | |
| WRZOMWDJOLIVQP-UHFFFAOYSA-N | |
| 5-chloro-2-methylaniline | |
| 7260 | |
| 141.6 | |
| 5-Chloro-2-methylaniline, 98% |
| 97.5 | |
| 237°C | |
| Authentic | |
| Glass bottle | |
| 1.5830 to 1.585 | |
| MFCD00007779 | |
| 12, 835 | |
| Solubility in water: < 1g/L (22°C). Other solubilities: soluble in alcohol,ether,benzene,chloroform and,dioxane | |
| CC1=CC=C(Cl)C=C1N | |
| 141.60 | |
| CHEBI:82422 | |
| 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
EINECSNumber : 202-452-6
RTECSNumber : XU5075000
TSCA : TSCA
Recommended Storage : Product may darken on storage
RUO â Research Use Only