missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$143.54 - $143.54
Chemical Identifiers
| CAS | 89694-48-4 |
|---|---|
| Molecular Formula | C7H8BClO3 |
| MDL Number | MFCD01318966 |
| InChI Key | FMBVAOHFMSQDGT-UHFFFAOYSA-N |
| PubChem CID | 2735751 |
| IUPAC Name | (5-chloro-2-methoxyphenyl)boronic acid |
Chemical Identifiers
| 89694-48-4 | |
| MFCD01318966 | |
| 2735751 |
| C7H8BClO3 | |
| FMBVAOHFMSQDGT-UHFFFAOYSA-N | |
| (5-chloro-2-methoxyphenyl)boronic acid |
Specifications
| 89694-48-4 | |
| White-Yellow | |
| MFCD01318966 | |
| 5-chloro-2-methoxyphenyl boronic acid, 5-chloro-2-methoxybenzeneboronic acid, 5-chloro-2-methoxyphenyl boranediol, 2-methoxy-5-chlorophenylboronic acid, 4-chloroanisole-2-boronic acid, boronic acid, 5-chloro-2-methoxyphenyl, 5-chloro-2-methoxyphenylboronicacid, pubchem1782, 5-chloro-2-methoxy-phenyl boronic acid | |
| COC1=C(C=C(Cl)C=C1)B(O)O | |
| 186.40 | |
| 186.40 | |
| 5-Chloro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride) |
| 145°C | |
| C7H8BClO3 | |
| 5 g | |
| FMBVAOHFMSQDGT-UHFFFAOYSA-N | |
| (5-chloro-2-methoxyphenyl)boronic acid | |
| 2735751 | |
| Crystalline Powder |
Safety and Handling
TSCA : No