missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxybenzenesulfonyl Chloride 95.0+%, TCI America™

Catalog No. p-7125625
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27405G 5 g
C274025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27405G Supplier TCI America Supplier No. C27405G
Only null left

Chemical Identifiers

CAS 22952-32-5
Molecular Formula C7H6Cl2O3S
Molecular Weight (g/mol) 241.082
MDL Number MFCD00052959
InChI Key FCJGLIMDVOTBLO-UHFFFAOYSA-N
Synonym 5-chloro-2-methoxybenzene-1-sulfonyl chloride, 5-chloro-2-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, 5-chloro-2-methoxy, chloro 5-chloro-2-methoxyphenyl sulfone, 5-chloro-2-methoxy-benzenesulfonyl chloride, acmc-1cq3l, ksc494k2b, 4-chloro-2-chlorosulphonyl anisole, 2-methoxy-5-chlorobenzenesulfonyl chloride, 5-chloro-2 methoxybenzenesulfonyl chloride
PubChem CID 2734272
IUPAC Name 5-chloro-2-methoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)Cl)S(=O)(=O)Cl

Specifications

Melting Point 101°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 241.08
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methoxybenzenesulfonyl Chloride