missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxybenzenesulfonyl chloride, 98%

Catalog No. p-7021852
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1436214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1436214 Supplier Thermo Scientific Chemicals Supplier No. A1436214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22952-32-5 | C7H6Cl2O3S | 241.082 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22952-32-5
Molecular Formula C7H6Cl2O3S
Molecular Weight (g/mol) 241.082
MDL Number MFCD00052959
InChI Key FCJGLIMDVOTBLO-UHFFFAOYSA-N
Synonym 5-chloro-2-methoxybenzene-1-sulfonyl chloride, 5-chloro-2-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, 5-chloro-2-methoxy, chloro 5-chloro-2-methoxyphenyl sulfone, 5-chloro-2-methoxy-benzenesulfonyl chloride, acmc-1cq3l, ksc494k2b, 4-chloro-2-chlorosulphonyl anisole, 2-methoxy-5-chlorobenzenesulfonyl chloride, 5-chloro-2 methoxybenzenesulfonyl chloride
PubChem CID 2734272
IUPAC Name 5-chloro-2-methoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)Cl)S(=O)(=O)Cl

Specifications

Melting Point 100°C to 104°C
Quantity 25 g
UN Number UN3096
Sensitivity Moisture sensitive
Formula Weight 241.1
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methoxybenzenesulfonyl chloride
Hazard Category H302+H312+H332-H314-H335-H500
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only