missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine, 97%, Thermo Scientific™

Catalog No. AAH3329203
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3329203 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3329203 Supplier Thermo Scientific Chemicals Supplier No. H3329203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in Suzuki, Stille, Sonogashira, carbonylation, Heck, Buchwald-Hartwig reaction.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1347814-95-2
Molecular Formula C8H6ClNO2
Molecular Weight (g/mol) 183.591
MDL Number MFCD20040113
InChI Key KEGRTUBZRMQESQ-UHFFFAOYSA-N
Synonym 5-chlorofuro 3,2-b pyridin-2-yl methanol, 5-chloro-2-hydroxymethyl furo 3,2-b pyridine
PubChem CID 73995151
IUPAC Name (5-chlorofuro[3,2-b]pyridin-2-yl)methanol
SMILES C1=CC(=NC2=C1OC(=C2)CO)Cl

Specifications

Quantity 1g
Formula Weight 183.59
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-(hydroxymethyl)furo[3,2-b]pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures