missing translation for 'onlineSavingsMsg'
Learn More

5'-Chloro-2'-hydroxy-3'-nitroacetophenone 98.0+%, TCI America™

Catalog No. p-7125753
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29735G 5 g
C297325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29735G Supplier TCI America Supplier No. C29735G
Only null left

Chemical Identifiers

CAS 84942-40-5
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.589
MDL Number MFCD00191921
InChI Key IUNBIQBAYUBIFD-UHFFFAOYSA-N
Synonym 5'-chloro-2'-hydroxy-3'-nitroacetophenone, 1-5-chloro-2-hydroxy-3-nitrophenyl ethanone, 2-acetyl-6-nitro-4-chlorophenol, 2-acetyl-6-nitro-4-chloro phenol, 2-acetyl-4-chloro-6-nitrophenol, 5-chloro-3-nitro-2-hydroxyacetophenone, benzoic acid,5-chloro-2-hydroxy-3-nitro, ethanone, 1-5-chloro-2-hydroxy-3-nitrophenyl, 1-acetyl-5-chloro-2-hydroxy-3-nitrobenzene, pubchem3355
PubChem CID 688237
SMILES CC(=O)C1=CC(=CC(=C1O)[N+](=O)[O-])Cl

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 5 g
Formula Weight 215.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Chloro-2′-hydroxy-3′-nitroacetophenone