missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-2'-deoxyuridine, MP Biomedicals™

Catalog No. MP210547891
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210547891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210547891 Supplier MP Biomedicals Inc Supplier No. 0210547891

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Readily incorporated, following phosphorylation, into newly synthesized DNA in place of thymidine. MP Biomedicals’ 5-chloro-2'-deoxyuridine (CldU) can be used as labeling substrate, thymidine analog, or to identify sites of DNA replication. Visible absorb

  • Used as a thymidine analog to study the miscoding potential of hypochlorous acid damage to DNA and DNA precursors
  • When used with antibody based immunofluorescent imaging, may be used in protocols to identify sites of DNA replication
  • May be used as a labeling substrate in conjunction with other halogenated uridine labeling substrates, such as bromodeoxyuridine (BrdU) and/or iododeoxyuridine (IdU)

Chemical Identifiers

CAS 50-90-8
Molecular Formula C9H11ClN2O5
Molecular Weight (g/mol) 262.646
InChI Key NJCXGFKPQSFZIB-XVMARJQXSA-N
Synonym 5-chloro-2'-deoxyuridine
PubChem CID 13831089
IUPAC Name 5-chloro-1-[(2S,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1C(C(OC1N2C=C(C(=O)NC2=O)Cl)CO)O

Specifications

Melting Point 166°C to 176°C
Quantity 1 g
Solubility Information Soluble in 1M NH4OH (20mg/1ml, colorless solution).
Formula Weight 262.6
Physical Form Powder
Recommended Storage Store at 0°C.