missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2'-deoxyuridine, MP Biomedicals™

Catalog No. ICN10547880
Change view
Click to view available options
Quantity:
0.1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10547880 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10547880 Supplier MP Biomedicals Inc Supplier No. 0210547880

Chemical Identifiers

CAS 50-90-8
Molecular Formula C9H11ClN2O5
Molecular Weight (g/mol) 262.646
InChI Key NJCXGFKPQSFZIB-XVMARJQXSA-N
Synonym 5-chloro-2'-deoxyuridine
PubChem CID 13831089
IUPAC Name 5-chloro-1-[(2S,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1C(C(OC1N2C=C(C(=O)NC2=O)Cl)CO)O

Specifications

Color White
Assay Percent Range >98%
Quantity 0.1 g
Formula Weight 262.65
Physical Form Powder
Chemical Name or Material 5-Chloro-2'-deoxyuridine