missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-cyanopyridine 98.0+%, TCI America™
$227.41 - $760.54
Chemical Identifiers
| CAS | 89809-64-3 |
|---|---|
| Molecular Formula | C6H3ClN2 |
| Molecular Weight (g/mol) | 138.55 |
| MDL Number | MFCD03788835 |
| InChI Key | WTHODOKFSYPTKA-UHFFFAOYSA-N |
| Synonym | 5-chloro-2-cyanopyridine, 5-chloropicolinonitrile, 3-chloro-6-cyanopyridine, 2-cyano-5-chloro pyridine, 2-pyridinecarbonitrile, 5-chloro, 2-cyano-5-chloropyridine, 6-cyano-3-chloropyridine, 5-chloro-2-pyridinecarbonitrile, pubchem2107, 5-chloropicolinenitrile |
| PubChem CID | 3833942 |
| IUPAC Name | 5-chloropyridine-2-carbonitrile |
| SMILES | ClC1=CN=C(C=C1)C#N |
Chemical Identifiers
| 89809-64-3 | |
| 138.55 | |
| WTHODOKFSYPTKA-UHFFFAOYSA-N | |
| 3833942 | |
| ClC1=CN=C(C=C1)C#N |
| C6H3ClN2 | |
| MFCD03788835 | |
| 5-chloro-2-cyanopyridine, 5-chloropicolinonitrile, 3-chloro-6-cyanopyridine, 2-cyano-5-chloro pyridine, 2-pyridinecarbonitrile, 5-chloro, 2-cyano-5-chloropyridine, 6-cyano-3-chloropyridine, 5-chloro-2-pyridinecarbonitrile, pubchem2107, 5-chloropicolinenitrile | |
| 5-chloropyridine-2-carbonitrile |
Specifications
| 89809-64-3 | |
| Yellow | |
| C6H3ClN2 | |
| 5 g | |
| 5-chloro-2-cyanopyridine, 5-chloropicolinonitrile, 3-chloro-6-cyanopyridine, 2-cyano-5-chloro pyridine, 2-pyridinecarbonitrile, 5-chloro, 2-cyano-5-chloropyridine, 6-cyano-3-chloropyridine, 5-chloro-2-pyridinecarbonitrile, pubchem2107, 5-chloropicolinenitrile | |
| ClC1=CN=C(C=C1)C#N | |
| 138.55 | |
| 138.55 | |
| Crystalline Powder |
| 109°C | |
| 110°C | |
| MFCD03788835 | |
| 3439 | |
| WTHODOKFSYPTKA-UHFFFAOYSA-N | |
| 5-chloropyridine-2-carbonitrile | |
| 3833942 | |
| ≥98.0% (GC) | |
| 5-Chloro-2-cyanopyridine |
Safety and Handling
TSCA : No