missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-cyanopyridine 98.0+%, TCI America™

Catalog No. C30235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30235G 5 g
C302325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30235G Supplier TCI America Supplier No. C30235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89809-64-3
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.55
MDL Number MFCD03788835
InChI Key WTHODOKFSYPTKA-UHFFFAOYSA-N
Synonym 5-chloro-2-cyanopyridine, 5-chloropicolinonitrile, 3-chloro-6-cyanopyridine, 2-cyano-5-chloro pyridine, 2-pyridinecarbonitrile, 5-chloro, 2-cyano-5-chloropyridine, 6-cyano-3-chloropyridine, 5-chloro-2-pyridinecarbonitrile, pubchem2107, 5-chloropicolinenitrile
PubChem CID 3833942
IUPAC Name 5-chloropyridine-2-carbonitrile
SMILES ClC1=CN=C(C=C1)C#N

Specifications

Melting Point 109°C
Color Yellow
Boiling Point 110°C
Quantity 5 g
UN Number 3439
Formula Weight 138.55
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-cyanopyridine