missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1H-pyrrolo[2,3-b]pyridine 98.0+%, TCI America™

Catalog No. p-7125769
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29961G 1 g
C29965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29961G Supplier TCI America Supplier No. C29961G

Chemical Identifiers

CAS 866546-07-8
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.581
MDL Number MFCD06659676
InChI Key MFZQJIKENSPRSJ-UHFFFAOYSA-N
Synonym 5-chloro-1h-pyrrolo 2,3-b pyridine, 5-chloro-7-azaindole, 1h-pyrrolo 2,3-b pyridine, 5-chloro, pubchem14704, ksc494e6r, 5-chloropyrrolo 2,3-b pyridine, 5-chloro-1h-pyrrolo 2,3-b pyridin
PubChem CID 24229213
IUPAC Name 5-chloro-1H-pyrrolo[2,3-b]pyridine
SMILES C1=CNC2=NC=C(C=C21)Cl

Specifications

Melting Point 165°C
Color Yellow
Quantity 1 g
Formula Weight 152.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-1H-pyrrolo[2,3-b]pyridine