missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methylimidazole 98.0+%, TCI America™

Catalog No. C23175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23175G 5 g
C231725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23175G Supplier TCI America Supplier No. C23175G

Chemical Identifiers

CAS 872-49-1
Molecular Formula C4H5ClN2
Molecular Weight (g/mol) 116.548
MDL Number MFCD00014505
InChI Key NYDGOZPYEABERA-UHFFFAOYSA-N
Synonym 5-chloro-1-methyl-1h-imidazole, 1-methyl-5-chloroimidazole, 1h-imidazole, 5-chloro-1-methyl, 5-chloro-1-methylimidazole, zlchem 661, pubchem7612, acmc-209qj7, 5-chloro-1-methyl imidazole, 5-chloro-1-methyl-imidazole, ksc494e7d
PubChem CID 70105
IUPAC Name 5-chloro-1-methylimidazole
SMILES CN1C=NC=C1Cl

Specifications

Color Yellow
Boiling Point 85°C
Quantity 5 g
Formula Weight 116.55
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Chloro-1-methylimidazole