missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-1-benzothiophene-3-carbonitrile, Thermo Scientific™

Catalog No. CC00066CB
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC00066CB Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC00066CB Supplier Thermo Scientific Chemicals Supplier No. CC00066CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 16296-79-0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H4ClNS
MDL Number MFCD02682022
Quantity 250mg
Synonym 5-chlorobenzo b thiophene-3-carbonitrile, benzo b thiophene-3-carbonitrile,5-chloro, 5-chlorothionaphthene-3-carbonitrile, 5-chloranyl-1-benzothiophene-3-carbonitrile, 5-chloro-1-benzo b thiophene-3-carbonitrile, benzo b thiophene-3-carbonitrile, 5-chloro
InChI Key SXWOPXJSURXXCM-UHFFFAOYSA-N
SMILES ClC1=CC=C2SC=C(C#N)C2=C1
IUPAC Name 5-chloro-1-benzothiophene-3-carbonitrile
Molecular Weight (g/mol) 193.65
PubChem CID 2776125
Formula Weight 193.66
Percent Purity 97%
Chemical Name or Material 5-Chloro-1-benzothiophene-3-carbonitrile
Show More Show Less