missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,3-benzodioxole, 98%

Catalog No. AAB2253409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2253409 10 g
AAB2253418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2253409 Supplier Thermo Scientific Chemicals Supplier No. B2253409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7228-38-8 | C7H5ClO2 | 156.565 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7228-38-8
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00010842
InChI Key ODQPZHOXLYATLC-UHFFFAOYSA-N
PubChem CID 138966
IUPAC Name 5-chloro-1,3-benzodioxole
SMILES C1OC2=C(O1)C=C(C=C2)Cl

Specifications

Density 1.34
Boiling Point 185°C to 187°C
Flash Point 93°C (201°F)
Refractive Index 1.556
Quantity 10 g
Synonym 5-chlorobenzo d 1,3 dioxole, 1-chloro-3,4-methylenedioxybenzene, 1,3-benzodioxole, 5-chloro, 5-chloro-2h-1,3-benzodioxole, 1,3-benzodioxole,5-chloro, 5-chloro-benzo 1,3 dioxole, pubchem3620, 4-chloro-1,2-methylenedioxybenzene, 5-chloro-1,3-benzodioxole, ukrorgsyn-bb bbr-008667
Formula Weight 156.57
Percent Purity 98%
Chemical Name or Material 5-Chloro-1,3-benzodioxole
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only