missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1,2,4-triazolo[4,3-a]pyridine, 97%

Catalog No. p-7055331
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63881MD 250 mg
AAH6388103 1 g
AAH6388106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63881MD Supplier Thermo Scientific Chemicals Supplier No. H63881MD
Only null left

CAS: 27187-13-9 | C6H4ClN3 | 153.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27187-13-9
Molecular Formula C6H4ClN3
Molecular Weight (g/mol) 153.57
MDL Number MFCD11109431
InChI Key FBQZXTMUYNKLRF-UHFFFAOYSA-N
Synonym 5-chloro-1,2,4 triazolo 4,3-a pyridine, 5-chloro-1,2,4-triazolo 4,3-a pyridine, 1,2,4-triazolo 4,3-a pyridine, 5-chloro, 5-chloro 1,2,4 triazolo 4,3-a pyridine
PubChem CID 324440
IUPAC Name 5-chloro-[1,2,4]triazolo[4,3-a]pyridine
SMILES ClC1=CC=CC2=NN=CN12

Specifications

Quantity 250 mg
Formula Weight 153.57
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Chloro-1,2,4-triazolo[4,3-a]pyridine
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only