missing translation for 'onlineSavingsMsg'
Learn More

5-Carboxyfluorescein N-Succinimidyl Ester 95.0+%, TCI America™

Catalog No. p-7125484
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C247920MG 20 mg
C2479100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C247920MG Supplier TCI America Supplier No. C247920MG
Only null left

Chemical Identifiers

CAS 92557-80-7
Molecular Formula C25H15NO9
Molecular Weight (g/mol) 473.393
MDL Number MFCD00077322
InChI Key GECIDMICWWDIBO-UHFFFAOYSA-N
PubChem CID 2762599
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylate
SMILES C1CC(=O)N(C1=O)OC(=O)C2=CC3=C(C=C2)C4(C5=C(C=C(C=C5)O)OC6=C4C=CC(=C6)O)OC3=O

Specifications

Quantity 20 mg
Formula Weight 473.39
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Carboxyfluorescein N-Succinimidyl Ester
TSCA No
Recommended Storage Freezer