missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Carbobenzoxyamino)valeric Acid 98.0+%, TCI America™
$432.46 - $1724.25
Chemical Identifiers
| CAS | 23135-50-4 |
|---|---|
| Molecular Formula | C13H17NO4 |
| Molecular Weight (g/mol) | 251.28 |
| MDL Number | MFCD00188458 |
| InChI Key | QYYPKLYDFCYGPG-UHFFFAOYSA-N |
| Synonym | 5-benzyloxy carbonyl amino pentanoic acid, 5-benzyloxycarbonylamino-pentanoic acid, 5-benzyloxycarbonylamino pentanoic acid, z-5-ava-oh, 5-carbobenzoxyamino valeric acid, 5-carbobenzoxyamino pentanoic acid, 5-phenylmethoxycarbonylamino pentanoic acid, 5-benzyloxycarbonylamino-pentanoicacid, pubchem20738 |
| PubChem CID | 3690024 |
| IUPAC Name | 5-{[(benzyloxy)carbonyl]amino}pentanoic acid |
| SMILES | OC(=O)CCCCNC(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 23135-50-4 | |
| 251.28 | |
| QYYPKLYDFCYGPG-UHFFFAOYSA-N | |
| 3690024 | |
| OC(=O)CCCCNC(=O)OCC1=CC=CC=C1 |
| C13H17NO4 | |
| MFCD00188458 | |
| 5-benzyloxy carbonyl amino pentanoic acid, 5-benzyloxycarbonylamino-pentanoic acid, 5-benzyloxycarbonylamino pentanoic acid, z-5-ava-oh, 5-carbobenzoxyamino valeric acid, 5-carbobenzoxyamino pentanoic acid, 5-phenylmethoxycarbonylamino pentanoic acid, 5-benzyloxycarbonylamino-pentanoicacid, pubchem20738 | |
| 5-{[(benzyloxy)carbonyl]amino}pentanoic acid |
Specifications
| 106°C | |
| White | |
| MFCD00188458 | |
| QYYPKLYDFCYGPG-UHFFFAOYSA-N | |
| 5-{[(benzyloxy)carbonyl]amino}pentanoic acid | |
| 251.28 | |
| 251.28 | |
| Crystalline Powder |
| 23135-50-4 | |
| C13H17NO4 | |
| 5-benzyloxy carbonyl amino pentanoic acid, 5-benzyloxycarbonylamino-pentanoic acid, 5-benzyloxycarbonylamino pentanoic acid, z-5-ava-oh, 5-carbobenzoxyamino valeric acid, 5-carbobenzoxyamino pentanoic acid, 5-phenylmethoxycarbonylamino pentanoic acid, 5-benzyloxycarbonylamino-pentanoicacid, pubchem20738 | |
| OC(=O)CCCCNC(=O)OCC1=CC=CC=C1 | |
| 5 g | |
| 3690024 | |
| ≥98.0% (T) | |
| 5-(Carbobenzoxyamino)valeric Acid |
Safety and Handling
TSCA : No