missing translation for 'onlineSavingsMsg'
Learn More

5-Carbamoylpyridine-3-boronic acid pinacol ester, 96%

Catalog No. AAH52330MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52330MD 250 mg
AAH5233003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52330MD Supplier Thermo Scientific Chemicals Supplier No. H52330MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1169402-51-0 | C12H17BN2O3 | 248.089 g/mol

5-Carbamoylpyridine-3-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Carbamoylpyridine-3-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1169402-51-0
Molecular Formula C12H17BN2O3
Molecular Weight (g/mol) 248.089
MDL Number MFCD11878274
InChI Key NJLZODHYCOZPBJ-UHFFFAOYSA-N
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nicotinamide, 5-carbamoylpyridine-3-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carboxamide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-nicotinamide, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl nicotinamide, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-3-carboxamide, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-pyridinecarboxamide
PubChem CID 59023117
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CN=C2)C(=O)N

Specifications

Quantity 250 mg
Formula Weight 248.09
Percent Purity 96%
Chemical Name or Material 5-Carbamoylpyridine-3-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only